Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF14
Entrez Gene ID
9928
Gene Name
kinesin family member 14
Gene Aliases
MKS12
Chromosome Location
Chr.1: 200551497 - 200620791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305792.1 NP_001292721.1 23 - 24 4101 148
NM_014875.2 NP_055690.1 25 - 26 4513 148
XM_011510231.2 XP_011508533.1 26 - 27 4607 148
XM_011510232.2 XP_011508534.1 25 - 26 4269 148
XM_011510233.2 XP_011508535.1 26 - 27 4400 148
XM_011510235.2 XP_011508537.1 26 - 27 4112 148
XM_011510236.1 XP_011508538.1 24 - 25 4396 148
XM_017003005.1 XP_016858494.1 24 - 25 4690 148
XM_017003006.1 XP_016858495.1 25 - 26 4298 148
XM_017003007.1 XP_016858496.1 25 - 26 3825 148
GenBank mRNA BC098582.1 - 15 - 16 2106 148
BC113742.1 - 24 - 25 4152 148
D26361.1 - 25 - 26 4513 148

View Full Product Details