Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
Chromosome Location
Chr.4: 88725954 - 89111398 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015045.2 NP_001015045.1 13 - 14 1849 83
NM_001265578.1 NP_001252507.1 12 - 13 1807 83
NM_001265579.1 NP_001252508.1 13 - 14 1849 83
NM_001265580.1 NP_001252509.1 11 - 12 1765 83
NM_014883.3 NP_055698.2 19 - 20 2701 83
XM_005262681.2 XP_005262738.1 18 - 19 2659 83
XM_005262682.2 XP_005262739.1 19 - 20 2800 83
XM_005262683.2 XP_005262740.1 19 - 20 2701 83
XM_005262684.3 XP_005262741.1 18 - 19 2480 83
XM_006714057.3 XP_006714120.1 16 - 17 1976 83
XM_011531516.1 XP_011529818.1 20 - 21 2722 83
XM_011531517.1 XP_011529819.1 18 - 19 2617 83
XM_011531518.1 XP_011529820.1 20 - 21 2851 83
XM_011531519.2 XP_011529821.1 18 - 19 2410 83
XM_017007624.1 XP_016863113.1 18 - 19 2659 83
XM_017007625.1 XP_016863114.1 18 - 19 2320 83
XM_017007626.1 XP_016863115.1 16 - 17 1954 83
XM_017007627.1 XP_016863116.1 18 - 19 2545 83
XM_017007628.1 XP_016863117.1 17 - 18 2438 83
XM_017007629.1 XP_016863118.1 19 - 20 4432 83
XM_017007630.1 XP_016863119.1 17 - 18 2345 83
XM_017007631.1 XP_016863120.1 18 - 19 4390 83
XM_017007632.1 XP_016863121.1 16 - 17 2284 83
XM_017007633.1 XP_016863122.1 18 - 19 4348 83
XM_017007634.1 XP_016863123.1 12 - 13 1758 83
GenBank mRNA AB020721.2 - 13 - 14 1566 83
AK027138.1 - 13 - 14 1689 83
AK091787.1 - 12 - 13 1967 83
AK296932.1 - 11 - 12 1634 83
BC086875.1 - 12 - 13 1648 83

View Full Product Details