Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADNP2
Entrez Gene ID
22850
Gene Name
ADNP homeobox 2
Gene Aliases
ZNF508
Chromosome Location
Chr.18: 80109031 - 80140346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014913.3 NP_055728.1 1 - 2 437 126
XM_011525883.2 XP_011524185.1 1 - 2 217 126
GenBank mRNA AB020670.1 - 1 - 2 167 126
AK292566.1 - 1 - 2 206 126
BC071589.1 - 1 - 2 437 126

View Full Product Details