The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014938.5 | NP_055753.3 | 9 - 10 | 1859 | 63 |
XM_006719290.3 | XP_006719353.1 | 9 - 10 | 1843 | 63 | |
XM_006719291.3 | XP_006719354.1 | 9 - 10 | 1843 | 63 | |
XM_006719292.3 | XP_006719355.1 | 9 - 10 | 1843 | 63 | |
XM_006719293.3 | XP_006719356.1 | 9 - 10 | 1843 | 63 | |
XM_006719294.3 | XP_006719357.1 | 3 - 4 | 902 | 63 | |
GenBank mRNA | AB020674.1 | - | 1 - 2 | 706 | 63 |
AF245480.1 | - | 1 - 2 | 706 | 63 | |
AF245481.1 | - | 1 - 2 | 706 | 63 | |
AF312918.1 | - | 9 - 10 | 1733 | 63 | |
BC069272.1 | - | 4 - 5 | 996 | 63 | |
BC151841.1 | - | 9 - 10 | 1841 | 63 |