The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286639.1 | NP_001273568.1 | 4 - 5 | 1470 | 61 |
NM_001286640.1 | NP_001273569.1 | 3 - 4 | 1312 | 61 | |
NM_014940.3 | NP_055755.1 | 5 - 6 | 1797 | 61 | |
GenBank mRNA | AB020679.1 | - | 5 - 6 | 1481 | 61 |
AF442486.1 | - | 4 - 5 | 1461 | 61 | |
AK023374.1 | - | 4 - 5 | 738 | 61 | |
AK023896.1 | - | 5 - 6 | 1525 | 61 | |
AK128411.1 | - | 2 - 3 | 1339 | 61 | |
AK293392.1 | - | 3 - 4 | 1032 | 61 | |
AK296467.1 | - | 4 - 5 | 1197 | 61 | |
BC024277.1 | - | 5 - 6 | 1777 | 61 |