Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB1
Entrez Gene ID
22890
Gene Name
zinc finger and BTB domain containing 1
Gene Aliases
ZNF909
Chromosome Location
Chr.14: 64504574 - 64533690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123329.1 NP_001116801.1 1 - 2 375 59
NM_014950.2 NP_055765.2 1 - 2 375 59
GenBank mRNA AB023214.1 - 1 - 2 246 59
AK291743.1 - 1 - 2 153 59
BX248777.1 - 1 - 2 375 59

View Full Product Details