Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPH3A
Entrez Gene ID
22895
Gene Name
rabphilin 3A
Gene Aliases
-
Chromosome Location
Chr.12: 112791538 - 112898881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143854.1 NP_001137326.1 16 - 17 1849 61
NM_014954.3 NP_055769.2 15 - 16 1837 61
XM_011538048.1 XP_011536350.1 16 - 17 1653 61
XM_017019034.1 XP_016874523.1 8 - 9 1107 61
GenBank mRNA AB023202.1 - 16 - 17 1770 61
AK296296.1 - 14 - 15 1388 61
AK316112.1 - 16 - 17 1684 61
BC002605.2 - 14 - 15 1575 61
BC009843.2 - 14 - 15 1498 61
BX647281.1 - 8 - 9 2443 61

View Full Product Details