The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037442.3 | NP_001032519.1 | 1 - 2 | 912 | 107 |
NM_014961.4 | NP_055776.1 | 1 - 2 | 912 | 107 | |
GenBank mRNA | AB020678.1 | - | 1 - 2 | 718 | 107 |
AK294421.1 | - | 1 - 2 | 97 | 107 | |
BC051716.1 | - | 1 - 2 | 780 | 107 | |
BC053837.1 | - | 1 - 2 | 742 | 107 | |
BC071988.1 | - | 1 - 2 | 715 | 107 | |
BC098433.1 | - | 1 - 2 | 742 | 107 |