Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD3
Entrez Gene ID
22903
Gene Name
BTB domain containing 3
Gene Aliases
dJ742J24.1
Chromosome Location
Chr.20: 11890723 - 11926595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282550.1 NP_001269479.1 2 - 3 319 76
NM_001282551.1 NP_001269480.1 2 - 3 415 76
NM_014962.3 NP_055777.1 1 - 2 737 76
NM_181443.3 NP_852108.2 2 - 3 500 76
XM_017027728.1 XP_016883217.1 3 - 4 648 76
XM_017027729.1 XP_016883218.1 3 - 4 580 76
GenBank mRNA AB023169.1 - 1 - 2 685 76
AK299281.1 - 2 - 3 500 76
BC093677.1 - 1 - 2 208 76
BC101527.1 - 1 - 2 207 76
BC109315.1 - 2 - 3 348 76
BC109316.1 - 2 - 3 348 76
CB961606.1 - 2 - 3 421 76
DC314595.1 - 2 - 3 415 76
DC418438.1 - 2 - 3 319 76

View Full Product Details