The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135608.1 | NP_001129080.1 | 11 - 12 | 1140 | 70 |
NM_015071.4 | NP_055886.1 | 11 - 12 | 1140 | 70 | |
XM_005268398.4 | XP_005268455.1 | 11 - 12 | 1377 | 70 | |
XM_005268399.4 | XP_005268456.1 | 11 - 12 | 1374 | 70 | |
XM_005268400.4 | XP_005268457.1 | 11 - 12 | 1375 | 70 | |
XM_005268402.4 | XP_005268459.1 | 11 - 12 | 1494 | 70 | |
XM_006714774.3 | XP_006714837.1 | 11 - 12 | 1381 | 70 | |
XM_011537610.2 | XP_011535912.1 | 11 - 12 | 1380 | 70 | |
XM_011537611.2 | XP_011535913.1 | 11 - 12 | 1380 | 70 | |
XM_017009247.1 | XP_016864736.1 | 11 - 12 | 1380 | 70 | |
XM_017009248.1 | XP_016864737.1 | 11 - 12 | 1376 | 70 | |
XM_017009249.1 | XP_016864738.1 | 11 - 12 | 1376 | 70 | |
XM_017009250.1 | XP_016864739.1 | 11 - 12 | 1376 | 70 | |
GenBank mRNA | AB014521.1 | - | 10 - 11 | 922 | 70 |
BC068555.1 | - | 11 - 12 | 1460 | 70 | |
KU178561.1 | - | 11 - 12 | 1105 | 70 | |
KU178562.1 | - | 11 - 12 | 1105 | 70 |