The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015517.4 | NP_056332.2 | 10 - 11 | 1382 | 77 |
NM_198971.2 | NP_945322.1 | 9 - 10 | 1265 | 77 | |
XM_011542744.2 | XP_011541046.1 | 9 - 10 | 1374 | 77 | |
XM_011542745.2 | XP_011541047.1 | 9 - 10 | 1224 | 77 | |
XM_011542746.1 | XP_011541048.1 | 8 - 9 | 1045 | 77 | |
XM_017017502.1 | XP_016872991.1 | 12 - 13 | 1653 | 77 | |
XM_017017503.1 | XP_016872992.1 | 10 - 11 | 1418 | 77 | |
XM_017017504.1 | XP_016872993.1 | 10 - 11 | 1293 | 77 | |
XM_017017505.1 | XP_016872994.1 | 10 - 11 | 1271 | 77 | |
XM_017017506.1 | XP_016872995.1 | 10 - 11 | 1234 | 77 | |
XM_017017507.1 | XP_016872996.1 | 12 - 13 | 1473 | 77 | |
XM_017017508.1 | XP_016872997.1 | 11 - 12 | 1348 | 77 | |
XM_017017509.1 | XP_016872998.1 | 9 - 10 | 1100 | 77 | |
GenBank mRNA | AK056362.1 | - | 9 - 10 | 1178 | 77 |
AK092869.1 | - | 4 - 5 | 1519 | 77 | |
AL080201.1 | - | 1 - 2 | 75 | 77 | |
BC001073.2 | - | 9 - 10 | 1236 | 77 | |
BC001945.1 | - | 9 - 10 | 1236 | 77 | |
BC012856.1 | - | 9 - 10 | 1177 | 77 | |
BC017234.2 | - | 9 - 10 | 1185 | 77 |