The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015542.3 | NP_056357.1 | 3 - 4 | 1255 | 65 |
NM_080599.2 | NP_542166.1 | 3 - 4 | 1425 | 65 | |
XM_011519449.2 | XP_011517751.1 | 3 - 4 | 1303 | 65 | |
GenBank mRNA | AB037829.1 | - | 2 - 3 | 1624 | 65 |
AF301013.1 | - | 3 - 4 | 1248 | 65 | |
AF318574.1 | - | 3 - 4 | 1225 | 65 | |
AK096191.1 | - | 4 - 5 | 1165 | 65 | |
AK310925.1 | - | 3 - 4 | 1214 | 65 | |
AY013249.1 | - | 2 - 3 | 1174 | 65 | |
BC114964.1 | - | 3 - 4 | 1219 | 65 | |
BC115737.1 | - | 3 - 4 | 1219 | 65 |