Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIPA1L1
Entrez Gene ID
26037
Gene Name
signal induced proliferation associated 1 like 1
Gene Aliases
E6TP1
Chromosome Location
Chr.14: 71320418 - 71741229 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284245.1 NP_001271174.1 10 - 11 3735 64
NM_001284246.1 NP_001271175.1 10 - 11 4324 64
NM_001284247.1 NP_001271176.1 9 - 10 3676 64
NM_015556.2 NP_056371.1 10 - 11 3735 64
XM_005267516.4 XP_005267573.1 10 - 11 4324 64
XM_006720111.2 XP_006720174.1 12 - 13 5614 64
XM_011536630.1 XP_011534932.1 14 - 15 4086 64
XM_011536631.1 XP_011534933.1 14 - 15 4497 64
XM_011536632.1 XP_011534934.1 13 - 14 4006 64
XM_011536635.2 XP_011534937.1 13 - 14 4153 64
XM_011536638.2 XP_011534940.1 9 - 10 2809 64
XM_017021159.1 XP_016876648.1 16 - 17 4243 64
XM_017021160.1 XP_016876649.1 14 - 15 4165 64
XM_017021161.1 XP_016876650.1 15 - 16 4118 64
XM_017021162.1 XP_016876651.1 17 - 18 4300 64
XM_017021163.1 XP_016876652.1 12 - 13 3967 64
XM_017021164.1 XP_016876653.1 12 - 13 3974 64
XM_017021165.1 XP_016876654.1 14 - 15 4043 64
XM_017021166.1 XP_016876655.1 15 - 16 4176 64
XM_017021167.1 XP_016876656.1 14 - 15 4051 64
XM_017021168.1 XP_016876657.1 14 - 15 4025 64
XM_017021169.1 XP_016876658.1 13 - 14 4178 64
XM_017021170.1 XP_016876659.1 13 - 14 4536 64
XM_017021171.1 XP_016876660.1 14 - 15 4109 64
XM_017021172.1 XP_016876661.1 13 - 14 3984 64
XM_017021173.1 XP_016876662.1 12 - 13 4111 64
XM_017021174.1 XP_016876663.1 12 - 13 4351 64
XM_017021175.1 XP_016876664.1 12 - 13 4469 64
XM_017021176.1 XP_016876665.1 17 - 18 4296 64
XM_017021177.1 XP_016876666.1 17 - 18 4285 64
XM_017021178.1 XP_016876667.1 15 - 16 4123 64
XM_017021179.1 XP_016876668.1 14 - 15 4026 64
XM_017021180.1 XP_016876669.1 18 - 19 4338 64
XM_017021181.1 XP_016876670.1 16 - 17 4243 64
XM_017021182.1 XP_016876671.1 15 - 16 4176 64
XM_017021183.1 XP_016876672.1 13 - 14 3984 64
XM_017021184.1 XP_016876673.1 17 - 18 4296 64
XM_017021185.1 XP_016876674.1 10 - 11 4324 64
XM_017021186.1 XP_016876675.1 16 - 17 4243 64
XM_017021187.1 XP_016876676.1 14 - 15 4109 64
XM_017021188.1 XP_016876677.1 13 - 14 3984 64
XM_017021189.1 XP_016876678.1 15 - 16 4093 64
XM_017021190.1 XP_016876679.1 16 - 17 4243 64
XM_017021191.1 XP_016876680.1 14 - 15 4051 64
XM_017021192.1 XP_016876681.1 13 - 14 3984 64
XM_017021193.1 XP_016876682.1 13 - 14 3982 64
XM_017021197.1 XP_016876686.1 15 - 16 2443 64
XM_017021198.1 XP_016876687.1 12 - 13 2184 64
XM_017021199.1 XP_016876688.1 16 - 17 2485 64
XM_017021200.1 XP_016876689.1 13 - 14 2251 64
GenBank mRNA AB007900.2 - 9 - 10 3459 64
AF090989.1 - 10 - 11 3722 64
AF090990.1 - 10 - 11 3722 64
AK122930.1 - 10 - 11 4324 64
AK302650.1 - 9 - 10 1974 64
BC146777.1 - 9 - 10 3459 64

View Full Product Details