The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015653.4 | NP_056468.3 | 4 - 5 | 876 | 121 |
XM_005261524.4 | XP_005261581.1 | 4 - 5 | 701 | 121 | |
XM_011530126.2 | XP_011528428.1 | 2 - 3 | 404 | 121 | |
GenBank mRNA | AK098586.1 | - | 4 - 5 | 876 | 121 |
AK225722.1 | - | 4 - 5 | 857 | 121 | |
AL442116.1 | - | 4 - 5 | 827 | 121 | |
BC003024.1 | - | 4 - 5 | 838 | 121 | |
BC009904.2 | - | 4 - 5 | 838 | 121 | |
CR456438.1 | - | 2 - 3 | 470 | 121 | |
CR457398.1 | - | 2 - 3 | 461 | 121 |