The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015718.2 | NP_056533.1 | 5 - 6 | 587 | 85 |
GenBank mRNA | AF190122.1 | - | 5 - 6 | 587 | 85 |
AF229177.1 | - | 3 - 4 | 326 | 85 |