The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003891.2 | NP_001003891.1 | 2 - 3 | 296 | 61 |
NM_001293234.1 | NP_001280163.1 | 2 - 3 | 296 | 61 | |
NM_001293235.1 | NP_001280164.1 | 2 - 3 | 285 | 61 | |
NM_001293236.1 | NP_001280165.1 | 2 - 3 | 296 | 61 | |
NM_001293237.1 | NP_001280166.1 | 2 - 3 | 488 | 61 | |
NM_015889.4 | NP_056973.2 | 2 - 3 | 296 | 61 | |
XM_006724263.3 | XP_006724326.1 | 2 - 3 | 254 | 61 | |
XM_011530214.2 | XP_011528516.1 | 3 - 4 | 340 | 61 | |
XM_011530218.2 | XP_011528520.1 | 2 - 3 | 296 | 61 | |
XM_011530219.1 | XP_011528521.1 | 2 - 3 | 296 | 61 | |
XM_011530220.2 | XP_011528522.1 | 2 - 3 | 296 | 61 | |
XM_017028821.1 | XP_016884310.1 | 2 - 3 | 296 | 61 | |
XM_017028822.1 | XP_016884311.1 | 2 - 3 | 296 | 61 | |
XM_017028823.1 | XP_016884312.1 | 1 - 2 | 91 | 61 | |
GenBank mRNA | AF056191.1 | - | 2 - 3 | 245 | 61 |
AF328769.1 | - | 2 - 3 | 159 | 61 | |
AK090465.1 | - | 2 - 3 | 228 | 61 | |
AK294540.1 | - | 2 - 3 | 304 | 61 | |
AK294734.1 | - | 2 - 3 | 484 | 61 | |
AK297789.1 | - | 2 - 3 | 222 | 61 | |
AK315540.1 | - | 2 - 3 | 256 | 61 | |
BC003078.1 | - | 2 - 3 | 217 | 61 | |
BC013985.2 | - | 2 - 3 | 199 | 61 | |
BX478424.1 | - | 2 - 3 | 239 | 61 | |
CB156495.1 | - | 2 - 3 | 488 | 61 | |
CR456537.1 | - | 2 - 3 | 197 | 61 | |
DA182752.1 | - | 2 - 3 | 299 | 61 |