Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHA8P1
Entrez Gene ID
51054
Gene Name
pleckstrin homology domain containing A8 pseudogene 1
Gene Aliases
PLEKHA9
Chromosome Location
Chr.12: 45173034 - 45216006 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_037144.1 2 - 3 259 79
GenBank mRNA AF103731.1 - 2 - 3 259 79

View Full Product Details