The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040455.1 | NP_001035545.1 | 1 - 2 | 228 | 82 |
GenBank mRNA | AF151799.1 | - | 1 - 2 | 270 | 82 |
AK292501.1 | - | 1 - 2 | 708 | 82 | |
AK292974.1 | - | 1 - 2 | 219 | 82 | |
AK303023.1 | - | 1 - 2 | 184 | 82 | |
AY358442.1 | - | 1 - 2 | 237 | 82 | |
BC114522.1 | - | 1 - 2 | 257 | 82 |