The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016107.3 | NP_057191.2 | 17 - 18 | 2917 | 80 |
GenBank mRNA | AF100742.1 | - | 7 - 8 | 912 | 80 |
AJ314790.1 | - | 17 - 18 | 2873 | 80 | |
AK025481.1 | - | 5 - 6 | 610 | 80 | |
AK123306.1 | - | 1 - 2 | 1313 | 80 | |
AK295297.1 | - | 4 - 5 | 463 | 80 | |
AK307808.1 | - | 3 - 4 | 255 | 80 | |
BC051893.1 | - | 10 - 11 | 1508 | 80 | |
BC062986.1 | - | 10 - 11 | 1433 | 80 | |
BC137084.1 | - | 17 - 18 | 2881 | 80 | |
BC137085.1 | - | 17 - 18 | 2881 | 80 |