The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040443.2 | NP_001035533.1 | 2 - 3 | 614 | 110 |
NM_001040444.2 | NP_001035534.1 | 2 - 3 | 290 | 110 | |
NR_135323.1 | 3 - 4 | 426 | 110 | ||
GenBank mRNA | AB011031.1 | - | 2 - 3 | 263 | 110 |
AF155105.1 | - | 2 - 3 | 233 | 110 | |
AK026228.1 | - | 2 - 3 | 254 | 110 | |
AK225986.1 | - | 3 - 4 | 376 | 110 | |
AK293242.1 | - | 2 - 3 | 614 | 110 | |
AK300410.1 | - | 2 - 3 | 254 | 110 | |
BC017212.2 | - | 2 - 3 | 243 | 110 | |
DB050568.1 | - | 3 - 4 | 381 | 110 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320727.1 | NP_001307656.1 | 12 - 13 | 2610 | 110 |
GenBank mRNA | JF729368.1 | - | 2 - 3 | 210 | 110 |
JF729370.1 | - | 2 - 3 | 225 | 110 |