The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016190.2 | NP_057274.1 | 2 - 3 | 214 | 81 |
GenBank mRNA | AF077831.1 | - | 2 - 3 | 192 | 81 |
AK291301.1 | - | 2 - 3 | 203 | 81 | |
AK298640.1 | - | 2 - 3 | 203 | 81 | |
AK316568.1 | - | 2 - 3 | 203 | 81 | |
BC030807.1 | - | 2 - 3 | 203 | 81 | |
DA420219.1 | - | 2 - 3 | 214 | 81 |