The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016298.3 | NP_057382.2 | 3 - 4 | 2126 | 97 |
GenBank mRNA | AB033021.1 | - | 3 - 4 | 2330 | 97 |
AF204674.1 | - | 3 - 4 | 2032 | 97 | |
AK314400.1 | - | 3 - 4 | 2048 | 97 | |
BC109275.1 | - | 2 - 3 | 1937 | 97 | |
BC109276.1 | - | 2 - 3 | 1937 | 97 |