The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317964.1 | NP_001304893.1 | 2 - 3 | 420 | 107 |
NM_016460.3 | NP_057544.2 | 2 - 3 | 420 | 107 | |
NM_020117.10 | NP_064502.9 | 2 - 3 | 420 | 107 | |
XM_011537656.2 | XP_011535958.1 | 2 - 3 | 286 | 107 | |
GenBank mRNA | AB037773.1 | - | 2 - 3 | 293 | 107 |
AF151026.1 | - | 2 - 3 | 260 | 107 | |
AK025441.1 | - | 2 - 3 | 256 | 107 | |
AK225372.1 | - | 1 - 2 | 118 | 107 | |
AK293750.1 | - | 2 - 3 | 250 | 107 | |
AK304134.1 | - | 2 - 3 | 236 | 107 | |
AK315178.1 | - | 2 - 3 | 269 | 107 | |
BC068041.1 | - | 2 - 3 | 240 | 107 | |
BC105581.1 | - | 2 - 3 | 262 | 107 | |
BC131798.1 | - | 2 - 3 | 158 | 107 | |
BC150213.1 | - | 2 - 3 | 271 | 107 | |
BC151214.1 | - | 2 - 3 | 221 | 107 | |
BC152422.1 | - | 2 - 3 | 293 | 107 | |
D84223.1 | - | 2 - 3 | 198 | 107 | |
DB086913.1 | - | 2 - 3 | 365 | 107 | |
DB091795.1 | - | 2 - 3 | 287 | 107 |