Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC174
Entrez Gene ID
51244
Gene Name
coiled-coil domain containing 174
Gene Aliases
C3orf19, HSPC212, IHPM, IHPMR
Chromosome Location
Chr.3: 14651683 - 14673186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006554.1 XP_016862043.1 7 - 8 876 61
GenBank mRNA AF151046.1 - 7 - 8 779 61

View Full Product Details