Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM7SF3
Entrez Gene ID
51768
Gene Name
transmembrane 7 superfamily member 3
Gene Aliases
-
Chromosome Location
Chr.12: 26966519 - 27014427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016551.2 NP_057635.1 9 - 10 1430 73
XM_011520690.2 XP_011518992.1 8 - 9 1293 73
XM_017019463.1 XP_016874952.1 9 - 10 1541 73
XM_017019464.1 XP_016874953.1 7 - 8 1103 73
GenBank mRNA AB032470.1 - 9 - 10 1229 73
AK002031.1 - 9 - 10 1196 73
AK023085.1 - 9 - 10 1418 73
BC005176.2 - 9 - 10 1355 73

View Full Product Details