Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECSIT
Entrez Gene ID
51295
Gene Name
ECSIT signalling integrator
Gene Aliases
SITPEC
Chromosome Location
Chr.19: 11505916 - 11529172 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142464.2 NP_001135936.1 2 - 3 234 95
NM_001243204.1 NP_001230133.1 2 - 3 244 95
NM_016581.4 NP_057665.2 2 - 3 234 95
GenBank mRNA AF243044.1 - 1 - 2 133 95
AK314905.1 - 2 - 3 244 95
BC000193.1 - 2 - 3 182 95
BC005119.1 - 2 - 3 177 95
BC008279.1 - 2 - 3 199 95
CR457204.1 - 1 - 2 100 95

View Full Product Details