The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100420.1 | NP_001093890.1 | 2 - 3 | 219 | 65 |
NM_001100421.1 | NP_001093891.1 | 2 - 3 | 219 | 65 | |
NM_017447.3 | NP_059143.3 | 2 - 3 | 219 | 65 | |
GenBank mRNA | AF239726.1 | - | 2 - 3 | 172 | 65 |
AK023825.1 | - | 2 - 3 | 184 | 65 | |
AK225123.1 | - | 2 - 3 | 190 | 65 | |
AK314471.1 | - | 2 - 3 | 192 | 65 | |
BC015468.2 | - | 1 - 2 | 135 | 65 | |
BF739854.1 | - | 2 - 3 | 218 | 65 | |
CR457317.1 | - | 1 - 2 | 128 | 65 | |
DA215855.1 | - | 2 - 3 | 192 | 65 |