The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145099.1 | NP_001138571.1 | 2 - 3 | 348 | 104 |
NM_017585.3 | NP_060055.2 | 2 - 3 | 348 | 104 | |
XM_011518189.2 | XP_011516491.2 | 2 - 3 | 321 | 104 | |
XM_017014236.1 | XP_016869725.1 | 2 - 3 | 330 | 104 | |
XM_017014237.1 | XP_016869726.1 | 2 - 3 | 321 | 104 | |
XM_017014238.1 | XP_016869727.1 | 2 - 3 | 321 | 104 | |
GenBank mRNA | AB209108.1 | - | 1 - 2 | 613 | 104 |
AJ011372.2 | - | 2 - 3 | 290 | 104 | |
AK074836.1 | - | 2 - 3 | 298 | 104 | |
AK074927.1 | - | 2 - 3 | 331 | 104 | |
AK096630.1 | - | 1 - 2 | 851 | 104 | |
AK222919.1 | - | 2 - 3 | 293 | 104 | |
AK225106.1 | - | 2 - 3 | 290 | 104 | |
AK294974.1 | - | 2 - 3 | 268 | 104 | |
BC013740.1 | - | 2 - 3 | 282 | 104 | |
DC305025.1 | - | 2 - 3 | 348 | 104 | |
Y17803.1 | - | 2 - 3 | 299 | 104 |