The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017589.3 | NP_060059.1 | 5 - 6 | 857 | 71 |
GenBank mRNA | AJ271351.1 | - | 4 - 5 | 662 | 71 |