Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
-
Chromosome Location
Chr.4: 168216291 - 168318807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017631.5 NP_060101.3 35 - 36 5115 68
XM_011532103.2 XP_011530405.1 35 - 36 5115 68
GenBank mRNA AK000042.1 - 7 - 8 916 68
AK026333.1 - 3 - 4 381 68
AK223184.1 - 7 - 8 916 68
AK225191.1 - 7 - 8 903 68
AL137464.1 - 2 - 3 301 68
BC020601.1 - 3 - 4 331 68
BC038115.2 - 35 - 36 5060 68
BC074781.2 - 7 - 8 894 68
BX648655.1 - 15 - 16 1957 68

View Full Product Details