Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDKN2AIP
Entrez Gene ID
55602
Gene Name
CDKN2A interacting protein
Gene Aliases
CARF
Chromosome Location
Chr.4: 183444591 - 183449064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017632.3 NP_060102.1 2 - 3 611 102
GenBank mRNA AF246705.1 - 2 - 3 559 102
AK000043.1 - 2 - 3 566 102
AK225848.1 - 2 - 3 424 102
BC022270.1 - 2 - 3 448 102

View Full Product Details