Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R9A
Entrez Gene ID
55607
Gene Name
protein phosphatase 1 regulatory subunit 9A
Gene Aliases
NRB1, NRBI, Neurabin-I
Chromosome Location
Chr.7: 94907202 - 95296415 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166160.1 NP_001159632.1 18 - 19 4007 83
NM_001166161.1 NP_001159633.1 16 - 17 3707 83
NM_001166163.1 NP_001159635.1 12 - 13 3089 83
XM_011516380.1 XP_011514682.1 19 - 20 4194 83
XM_011516381.1 XP_011514683.1 19 - 20 4070 83
XM_011516382.2 XP_011514684.1 19 - 20 4083 83
XM_011516383.1 XP_011514685.1 19 - 20 4124 83
XM_011516389.1 XP_011514691.1 17 - 18 3941 83
XM_017012394.1 XP_016867883.1 19 - 20 4016 83
XM_017012397.1 XP_016867886.1 19 - 20 3956 83
XM_017012398.1 XP_016867887.1 17 - 18 3953 83
XM_017012401.1 XP_016867890.1 18 - 19 3839 83
XM_017012403.1 XP_016867892.1 17 - 18 3773 83
XM_017012407.1 XP_016867896.1 13 - 14 3155 83
XM_017012408.1 XP_016867897.1 17 - 18 2285 83
XM_017012410.1 XP_016867899.1 18 - 19 4073 83
XM_017012411.1 XP_016867900.1 12 - 13 6687 83
GenBank mRNA AK000075.1 - 6 - 7 838 83
AK225204.1 - 6 - 7 838 83
BC144108.1 - 16 - 17 3524 83
BC144109.1 - 12 - 13 2904 83
BC150636.1 - 17 - 18 3955 83

View Full Product Details