The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145798.2 | NP_665741.1 | 7 - 8 | 800 | 72 |
XM_017024122.1 | XP_016879611.1 | 7 - 8 | 822 | 72 | |
XM_017024123.1 | XP_016879612.1 | 7 - 8 | 940 | 72 | |
XM_017024124.1 | XP_016879613.1 | 6 - 7 | 787 | 72 | |
GenBank mRNA | AF323729.1 | - | 7 - 8 | 789 | 72 |
AF392446.1 | - | 7 - 8 | 853 | 72 | |
AK289523.1 | - | 7 - 8 | 822 | 72 | |
BC028010.1 | - | 7 - 8 | 799 | 72 | |
BC065482.1 | - | 7 - 8 | 772 | 72 |