Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44418251-44418744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNF4A
Entrez Gene ID
3172
Gene Name
hepatocyte nuclear factor 4 alpha
Gene Aliases
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14
UniGene
Hs.116462
Gene Chromosome Location
Chr.20: 44355801-44432845 on Build GRCh38
Transcripts
NM_000457.4 NM_001030003.2 NM_001030004.2 NM_001258355.1 NM_001287182.1 NM_001287183.1 NM_001287184.1 NM_175914.4 NM_178849.2 NM_178850.2 XM_005260407.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5581319 c.610G>A p.E204K Chr.20: 44418452-44418452 on Build GRCh38
7401253 c.652G>A p.A218T Chr.20: 44418428-44418428 on Build GRCh38
4802163 c.687G>T p.L229L Chr.20: 44418463-44418463 on Build GRCh38
6226349 c.612G>A p.E204E Chr.20: 44418454-44418454 on Build GRCh38
5675718 c.681C>G p.H227Q Chr.20: 44418457-44418457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs193922476 Chr.20: 44418502-44418504 on Build GRCh38
rs907580369 Chr.20: 44418596-44418596 on Build GRCh38
rs944488107 Chr.20: 44418603-44418603 on Build GRCh38
rs756051164 Chr.20: 44418621-44418621 on Build GRCh38
rs754623231 Chr.20: 44418475-44418475 on Build GRCh38
rs767221741 Chr.20: 44418561-44418561 on Build GRCh38
rs778308750 Chr.20: 44418544-44418544 on Build GRCh38
rs989471061 Chr.20: 44418505-44418505 on Build GRCh38
rs773632165 Chr.20: 44418534-44418534 on Build GRCh38
rs753476712 Chr.20: 44418452-44418452 on Build GRCh38
rs771999909 Chr.20: 44418547-44418548 on Build GRCh38
rs376586083 Chr.20: 44418499-44418499 on Build GRCh38
rs772349653 Chr.20: 44418503-44418503 on Build GRCh38
rs746375200 Chr.20: 44418700-44418700 on Build GRCh38
rs930466800 Chr.20: 44418572-44418572 on Build GRCh38
rs759084238 Chr.20: 44418466-44418466 on Build GRCh38
rs930293788 Chr.20: 44418352-44418352 on Build GRCh38
rs980107988 Chr.20: 44418711-44418711 on Build GRCh38
rs752171449 Chr.20: 44418562-44418562 on Build GRCh38
rs550636705 Chr.20: 44418331-44418331 on Build GRCh38
rs555086332 Chr.20: 44418609-44418609 on Build GRCh38
rs376544046 Chr.20: 44418519-44418519 on Build GRCh38
rs779187999 Chr.20: 44418683-44418683 on Build GRCh38
rs1047948629 Chr.20: 44418367-44418367 on Build GRCh38
rs775641733 Chr.20: 44418406-44418406 on Build GRCh38
rs777727657 Chr.20: 44418498-44418498 on Build GRCh38
rs930324423 Chr.20: 44418620-44418620 on Build GRCh38
rs747120718 Chr.20: 44418518-44418518 on Build GRCh38
rs1050001441 Chr.20: 44418628-44418628 on Build GRCh38
rs202105574 Chr.20: 44418500-44418500 on Build GRCh38
rs1000406853 Chr.20: 44418427-44418427 on Build GRCh38
rs6103731 Chr.20: 44418653-44418653 on Build GRCh38
rs751211080 Chr.20: 44418530-44418530 on Build GRCh38
rs766995199 Chr.20: 44418601-44418601 on Build GRCh38
rs995864334 Chr.20: 44418623-44418623 on Build GRCh38
rs1014280636 Chr.20: 44418661-44418661 on Build GRCh38
rs772107666 Chr.20: 44418400-44418400 on Build GRCh38
rs139591750 Chr.20: 44418511-44418511 on Build GRCh38
rs759052426 Chr.20: 44418552-44418552 on Build GRCh38
rs995980303 Chr.20: 44418651-44418651 on Build GRCh38
rs920552324 Chr.20: 44418538-44418538 on Build GRCh38
rs11086925 Chr.20: 44418708-44418708 on Build GRCh38
rs928135261 Chr.20: 44418581-44418581 on Build GRCh38
rs928251260 Chr.20: 44418283-44418283 on Build GRCh38
rs554777680 Chr.20: 44418590-44418590 on Build GRCh38
rs1032164393 Chr.20: 44418429-44418429 on Build GRCh38
rs748089076 Chr.20: 44418545-44418546 on Build GRCh38
rs1040067153 Chr.20: 44418311-44418311 on Build GRCh38
rs770124939 Chr.20: 44418529-44418529 on Build GRCh38
rs542275075 Chr.20: 44418688-44418688 on Build GRCh38
rs749046396 Chr.20: 44418639-44418639 on Build GRCh38
rs3212200 Chr.20: 44418289-44418289 on Build GRCh38
rs766033602 Chr.20: 44418355-44418355 on Build GRCh38
rs140446007 Chr.20: 44418348-44418348 on Build GRCh38
rs762312899 Chr.20: 44418440-44418440 on Build GRCh38
rs761201175 Chr.20: 44418409-44418409 on Build GRCh38
rs1000737158 Chr.20: 44418638-44418638 on Build GRCh38
rs777099767 Chr.20: 44418436-44418436 on Build GRCh38
rs113495257 Chr.20: 44418398-44418398 on Build GRCh38
rs893033040 Chr.20: 44418660-44418660 on Build GRCh38
rs372375706 Chr.20: 44418526-44418526 on Build GRCh38
rs752216751 Chr.20: 44418489-44418489 on Build GRCh38
rs576083592 Chr.20: 44418634-44418634 on Build GRCh38
rs779051589 Chr.20: 44418385-44418385 on Build GRCh38
rs751231466 Chr.20: 44418451-44418451 on Build GRCh38
rs1054760058 Chr.20: 44418301-44418301 on Build GRCh38
rs886221033 Chr.20: 44418626-44418626 on Build GRCh38
rs755998338 Chr.20: 44418493-44418493 on Build GRCh38
rs369710660 Chr.20: 44418520-44418520 on Build GRCh38
rs766035524 Chr.20: 44418442-44418442 on Build GRCh38
rs568820315 Chr.20: 44418612-44418612 on Build GRCh38
rs566659632 Chr.20: 44418570-44418570 on Build GRCh38
rs771153594 Chr.20: 44418384-44418384 on Build GRCh38
rs769305871 Chr.20: 44418414-44418414 on Build GRCh38

View Full Product Details