Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44419479-44419982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNF4A
Entrez Gene ID
3172
Gene Name
hepatocyte nuclear factor 4 alpha
Gene Aliases
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14
UniGene
Hs.116462
Gene Chromosome Location
Chr.20: 44355801-44432845 on Build GRCh38
Transcripts
NM_000457.4 NM_001030003.2 NM_001030004.2 NM_001258355.1 NM_001287182.1 NM_001287183.1 NM_001287184.1 NM_175914.4 NM_178849.2 NM_178850.2 XM_005260407.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4690811 c.702G>A p.P234P Chr.20: 44419752-44419752 on Build GRCh38
5631896 c.703G>A p.E235K Chr.20: 44419753-44419753 on Build GRCh38
3292818 c.745G>T p.E249* Chr.20: 44419795-44419795 on Build GRCh38
4953254 c.803T>G p.I268S Chr.20: 44419787-44419787 on Build GRCh38
4806249 c.880T>A p.F294I Chr.20: 44419864-44419864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776127911 Chr.20: 44419761-44419761 on Build GRCh38
rs73111879 Chr.20: 44419611-44419611 on Build GRCh38
rs997659238 Chr.20: 44419949-44419949 on Build GRCh38
rs748714111 Chr.20: 44419841-44419841 on Build GRCh38
rs372639360 Chr.20: 44419773-44419773 on Build GRCh38
rs775178928 Chr.20: 44419748-44419748 on Build GRCh38
rs193922477 Chr.20: 44419818-44419818 on Build GRCh38
rs748284293 Chr.20: 44419811-44419811 on Build GRCh38
rs6093979 Chr.20: 44419845-44419845 on Build GRCh38
rs765066730 Chr.20: 44419887-44419887 on Build GRCh38
rs756539709 Chr.20: 44419817-44419817 on Build GRCh38
rs770348231 Chr.20: 44419724-44419724 on Build GRCh38
rs866652936 Chr.20: 44419952-44419952 on Build GRCh38
rs750252688 Chr.20: 44419687-44419687 on Build GRCh38
rs913445184 Chr.20: 44419535-44419535 on Build GRCh38
rs928584332 Chr.20: 44419526-44419526 on Build GRCh38
rs1005078946 Chr.20: 44419591-44419591 on Build GRCh38
rs150078978 Chr.20: 44419746-44419746 on Build GRCh38
rs750234619 Chr.20: 44419533-44419533 on Build GRCh38
rs927455177 Chr.20: 44419566-44419566 on Build GRCh38
rs1004852897 Chr.20: 44419641-44419641 on Build GRCh38
rs145280017 Chr.20: 44419776-44419776 on Build GRCh38
rs773386088 Chr.20: 44419955-44419955 on Build GRCh38
rs1045844659 Chr.20: 44419550-44419550 on Build GRCh38
rs11574742 Chr.20: 44419506-44419506 on Build GRCh38
rs185240889 Chr.20: 44419679-44419679 on Build GRCh38
rs917593998 Chr.20: 44419833-44419833 on Build GRCh38
rs6031592 Chr.20: 44419794-44419794 on Build GRCh38
rs997772048 Chr.20: 44419625-44419625 on Build GRCh38
rs775961355 Chr.20: 44419853-44419853 on Build GRCh38
rs556013419 Chr.20: 44419521-44419521 on Build GRCh38
rs141943140 Chr.20: 44419830-44419830 on Build GRCh38
rs746602886 Chr.20: 44419844-44419844 on Build GRCh38
rs990201626 Chr.20: 44419669-44419669 on Build GRCh38
rs780467098 Chr.20: 44419692-44419692 on Build GRCh38
rs1049704009 Chr.20: 44419942-44419942 on Build GRCh38
rs564114816 Chr.20: 44419674-44419674 on Build GRCh38
rs199624185 Chr.20: 44419922-44419922 on Build GRCh38
rs6093980 Chr.20: 44419849-44419849 on Build GRCh38
rs764880264 Chr.20: 44419686-44419686 on Build GRCh38
rs781764367 Chr.20: 44419709-44419709 on Build GRCh38
rs757219506 Chr.20: 44419684-44419684 on Build GRCh38
rs781364316 Chr.20: 44419795-44419795 on Build GRCh38
rs758507396 Chr.20: 44419537-44419537 on Build GRCh38
rs137853334 Chr.20: 44419813-44419813 on Build GRCh38
rs145542196 Chr.20: 44419760-44419760 on Build GRCh38
rs141585334 Chr.20: 44419923-44419923 on Build GRCh38
rs1049163207 Chr.20: 44419575-44419575 on Build GRCh38
rs773402510 Chr.20: 44419732-44419732 on Build GRCh38
rs745649432 Chr.20: 44419699-44419699 on Build GRCh38
rs755171500 Chr.20: 44419698-44419698 on Build GRCh38
rs778511916 Chr.20: 44419532-44419532 on Build GRCh38
rs541400263 Chr.20: 44419645-44419645 on Build GRCh38
rs950319956 Chr.20: 44419542-44419542 on Build GRCh38
rs531607662 Chr.20: 44419882-44419882 on Build GRCh38
rs369816227 Chr.20: 44419889-44419889 on Build GRCh38
rs760173369 Chr.20: 44419752-44419752 on Build GRCh38
rs1028937748 Chr.20: 44419767-44419767 on Build GRCh38
rs748537070 Chr.20: 44419722-44419722 on Build GRCh38
rs937532583 Chr.20: 44419577-44419577 on Build GRCh38
rs758505661 Chr.20: 44419689-44419689 on Build GRCh38
rs141620976 Chr.20: 44419633-44419633 on Build GRCh38
rs902133525 Chr.20: 44419624-44419624 on Build GRCh38
rs747065787 Chr.20: 44419694-44419694 on Build GRCh38
rs761602960 Chr.20: 44419771-44419771 on Build GRCh38
rs1032055611 Chr.20: 44419784-44419784 on Build GRCh38
rs6093981 Chr.20: 44419893-44419893 on Build GRCh38
rs139779712 Chr.20: 44419774-44419774 on Build GRCh38
rs200985945 Chr.20: 44419772-44419772 on Build GRCh38
rs193922478 Chr.20: 44419906-44419907 on Build GRCh38
rs577733353 Chr.20: 44419677-44419677 on Build GRCh38
rs879833108 Chr.20: 44419654-44419654 on Build GRCh38
rs770348557 Chr.20: 44419927-44419927 on Build GRCh38
rs768316428 Chr.20: 44419847-44419847 on Build GRCh38
rs376013528 Chr.20: 44419741-44419741 on Build GRCh38

View Full Product Details