The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193455.1 | NP_001180384.1 | 6 - 7 | 1092 | 123 |
NM_017755.5 | NP_060225.4 | 7 - 8 | 1197 | 123 | |
NR_037947.1 | 6 - 7 | 1112 | 123 | ||
GenBank mRNA | AB255451.1 | - | - | 1196 | 123 |
AK000310.1 | - | 1 - 2 | 193 | 123 | |
AK055456.1 | - | 6 - 7 | 1042 | 123 | |
AK291144.1 | - | 7 - 8 | 880 | 123 | |
AK298980.1 | - | 6 - 7 | 774 | 123 | |
BC001041.2 | - | 5 - 6 | 553 | 123 | |
BC137083.1 | - | 7 - 8 | 878 | 123 |