The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040125.1 | NP_001035214.1 | 1 - 2 | 645 | 75 |
NM_017765.2 | NP_060235.2 | 2 - 3 | 275 | 75 | |
NR_109848.1 | 2 - 3 | 275 | 75 | ||
XM_006710710.2 | XP_006710773.1 | 2 - 3 | 199 | 75 | |
XM_011541634.2 | XP_011539936.1 | 2 - 3 | 227 | 75 | |
GenBank mRNA | AK000327.1 | - | 2 - 3 | 195 | 75 |
AK125580.1 | - | 1 - 2 | 563 | 75 | |
AK130171.1 | - | 2 - 3 | 193 | 75 | |
AK130175.1 | - | 2 - 3 | 195 | 75 | |
DA202894.1 | - | 2 - 3 | 275 | 75 |