The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017801.2 | NP_060271.1 | 3 - 4 | 585 | 120 |
GenBank mRNA | AF479261.1 | - | 3 - 4 | 1082 | 120 |
AK000403.1 | - | 3 - 4 | 526 | 120 | |
BC002797.2 | - | 3 - 4 | 541 | 120 | |
CR457231.1 | - | 3 - 4 | 419 | 120 |