The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017826.2 | NP_060296.2 | 9 - 10 | 1089 | 72 |
GenBank mRNA | AK000456.1 | - | 9 - 10 | 1013 | 72 |
AY884305.1 | - | 8 - 9 | 931 | 72 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198910.1 | NP_001185839.1 | 14 - 15 | 1546 | 72 |
GenBank mRNA | AK301863.1 | - | 14 - 15 | 1533 | 72 |