Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TUG1
Entrez Gene ID
55000
Gene Name
taurine up-regulated 1 (non-protein coding)
Gene Aliases
LINC00080, NCRNA00080, TI-227H
Chromosome Location
Chr.22: 30969211 - 30979395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002323.2 1 - 2 2572 70
NR_110493.1 1 - 2 2572 70

View Full Product Details