Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIM1L
Entrez Gene ID
55057
Gene Name
absent in melanoma 1-like
Gene Aliases
CRYBG2
Chromosome Location
Chr.1: 26321859 - 26354130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039775.3 NP_001034864.2 1 - 2 99 66
GenBank mRNA AK000902.1 - 1 - 2 99 66

View Full Product Details