The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018013.3 | NP_060483.3 | 6 - 7 | 3133 | 76 |
XM_005267041.4 | XP_005267098.1 | 7 - 8 | 3294 | 76 | |
XM_005267042.4 | XP_005267099.1 | 6 - 7 | 3199 | 76 | |
XM_011535921.2 | XP_011534223.1 | 7 - 8 | 2818 | 76 | |
XM_011535923.2 | XP_011534225.1 | 2 - 3 | 2032 | 76 | |
XM_017010991.1 | XP_016866480.1 | 5 - 6 | 3656 | 76 | |
XM_017010992.1 | XP_016866481.1 | 5 - 6 | 2425 | 76 | |
XM_017010993.1 | XP_016866482.1 | 5 - 6 | 2455 | 76 | |
XM_017010994.1 | XP_016866483.1 | 5 - 6 | 2414 | 76 | |
GenBank mRNA | BC113966.1 | - | 2 - 3 | 426 | 76 |