The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143824.1 | NP_001137296.1 | 5 - 6 | 608 | 107 |
NM_018018.4 | NP_060488.2 | 6 - 7 | 800 | 107 | |
XM_005268997.2 | XP_005269054.1 | 5 - 6 | 609 | 107 | |
GenBank mRNA | AB055003.1 | - | 4 - 5 | 432 | 107 |
AF193836.1 | - | 5 - 6 | 588 | 107 | |
AF305814.1 | - | 6 - 7 | 765 | 107 | |
AK001053.1 | - | 6 - 7 | 676 | 107 | |
AK291168.1 | - | 5 - 6 | 609 | 107 | |
BC069819.1 | - | 5 - 6 | 661 | 107 | |
BC101827.1 | - | 4 - 5 | 445 | 107 | |
BC104913.1 | - | 4 - 5 | 444 | 107 |