The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168347.2 | NP_001161819.1 | 4 - 5 | 581 | 96 |
NM_001168348.2 | NP_001161820.1 | 4 - 5 | 467 | 96 | |
NM_018044.4 | NP_060514.1 | 4 - 5 | 581 | 96 | |
NM_148956.3 | NP_683759.1 | 4 - 5 | 581 | 96 | |
XM_005277604.4 | XP_005277661.1 | 4 - 5 | 558 | 96 | |
GenBank mRNA | AF412028.1 | - | 4 - 5 | 520 | 96 |
AF420249.1 | - | 4 - 5 | 512 | 96 | |
AK001129.1 | - | 4 - 5 | 520 | 96 | |
AK098619.1 | - | 4 - 5 | 540 | 96 | |
AK125667.1 | - | 3 - 4 | 1203 | 96 | |
AK126375.1 | - | 4 - 5 | 404 | 96 | |
AK298221.1 | - | 4 - 5 | 530 | 96 | |
AK315361.1 | - | 4 - 5 | 522 | 96 | |
BC008084.2 | - | 4 - 5 | 521 | 96 |