Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ARHGAP17
- Entrez Gene ID
-
55114
- Gene Name
- Rho GTPase activating protein 17
- Gene Aliases
-
MST066, MST110, MSTP038, MSTP066, MSTP110, NADRIN, PP367, PP4534, RICH-1, RICH1, WBP15
- Chromosome Location
-
Chr.16: 24919384 - 25015601 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK001170.1 |
- |
1 - 2 |
613 |
84 |
View Full Product Details