The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018110.3 | NP_060580.2 | 2 - 3 | 416 | 78 |
XM_005256042.3 | XP_005256099.1 | 1 - 2 | 358 | 78 | |
XM_005256043.2 | XP_005256100.1 | 1 - 2 | 358 | 78 | |
GenBank mRNA | AF466369.1 | - | 1 - 2 | 66 | 78 |
AK001350.1 | - | 2 - 3 | 343 | 78 | |
AK307270.1 | - | 1 - 2 | 315 | 78 | |
BC001540.2 | - | 1 - 2 | 115 | 78 | |
BC003541.1 | - | 2 - 3 | 358 | 78 |