The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165962.1 | NP_001159434.1 | 13 - 14 | 1306 | 96 |
NM_018127.6 | NP_060597.4 | 14 - 15 | 1426 | 96 | |
NM_173717.1 | NP_776065.1 | 14 - 15 | 1423 | 96 | |
GenBank mRNA | AF304370.1 | - | 14 - 15 | 1309 | 96 |
AK001392.1 | - | 14 - 15 | 1380 | 96 | |
AK074244.1 | - | 6 - 7 | 548 | 96 | |
AK094333.1 | - | 6 - 7 | 1137 | 96 | |
AK124838.1 | - | 3 - 4 | 855 | 96 | |
AK125030.1 | - | 13 - 14 | 1571 | 96 | |
AK298397.1 | - | 13 - 14 | 1241 | 96 | |
AK300011.1 | - | 14 - 15 | 1301 | 96 | |
AK304589.1 | - | 9 - 10 | 708 | 96 | |
BC001939.1 | - | 14 - 15 | 1369 | 96 | |
BC004158.2 | - | 14 - 15 | 1369 | 96 | |
CR457261.1 | - | 14 - 15 | 1309 | 96 | |
KU178737.1 | - | 14 - 15 | 1309 | 96 |