Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SETD5
Entrez Gene ID
55209
Gene Name
SET domain containing 5
Gene Aliases
-
Chromosome Location
Chr.3: 9397700 - 9478154 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080517.2 NP_001073986.1 16 - 17 2801 129
NM_001292043.1 NP_001278972.1 18 - 19 3065 129
XM_005265301.1 XP_005265358.1 16 - 17 2839 129
XM_005265303.1 XP_005265360.1 18 - 19 3220 129
XM_011533920.1 XP_011532222.1 19 - 20 3315 129
XM_011533921.1 XP_011532223.1 18 - 19 2917 129
XM_011533922.1 XP_011532224.1 17 - 18 2878 129
XM_011533927.1 XP_011532229.1 19 - 20 3258 129
XM_011533928.1 XP_011532230.1 17 - 18 2821 129
XM_011533930.1 XP_011532232.1 19 - 20 3259 129
XM_011533932.1 XP_011532234.1 17 - 18 3026 129
XM_011533933.1 XP_011532235.1 15 - 16 2395 129
XM_017006767.1 XP_016862256.1 19 - 20 4977 129
XM_017006768.1 XP_016862257.1 17 - 18 4491 129
XM_017006769.1 XP_016862258.1 17 - 18 6594 129
XM_017006770.1 XP_016862259.1 18 - 19 6633 129
XM_017006771.1 XP_016862260.1 16 - 17 2839 129
XM_017006772.1 XP_016862261.1 17 - 18 6594 129
XM_017006773.1 XP_016862262.1 17 - 18 6537 129
XM_017006774.1 XP_016862263.1 16 - 17 2782 129
XM_017006775.1 XP_016862264.1 17 - 18 6537 129
XM_017006776.1 XP_016862265.1 18 - 19 3122 129
XM_017006777.1 XP_016862266.1 18 - 19 3065 129
XM_017006778.1 XP_016862267.1 19 - 20 3106 129
XM_017006779.1 XP_016862268.1 17 - 18 3083 129
XM_017006780.1 XP_016862269.1 18 - 19 3067 129
XM_017006781.1 XP_016862270.1 19 - 20 3106 129
XM_017006782.1 XP_016862271.1 18 - 19 6633 129
XM_017006783.1 XP_016862272.1 20 - 21 3331 129
XM_017006784.1 XP_016862273.1 18 - 19 6633 129
GenBank mRNA AB051544.1 - 12 - 13 2595 129
AK001569.1 - 5 - 6 129
AK024384.1 - 4 - 5 895 129
AK091393.1 - 13 - 14 2104 129
AK092046.1 - 18 - 19 3020 129
AK127298.1 - 6 - 7 1688 129
AK289529.1 - 7 - 8 1383 129
AL442073.1 - 3 - 4 781 129
BC020956.1 - 5 - 6 1015 129
BX648380.1 - 8 - 9 1403 129
CR627408.1 - 15 - 16 2417 129

View Full Product Details