Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D13
Entrez Gene ID
54662
Gene Name
TBC1 domain family member 13
Gene Aliases
-
Chromosome Location
Chr.9: 128787207 - 128810432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286772.1 NP_001273701.1 2 - 3 243 95
NM_018201.4 NP_060671.3 2 - 3 243 95
GenBank mRNA AB449899.1 - 2 - 3 96 95
AK001605.1 - 2 - 3 101 95
AK289777.1 - 2 - 3 155 95
BC014898.1 - 2 - 3 118 95
BC132817.1 - 2 - 3 116 95
BC136509.1 - 2 - 3 116 95
BC150311.1 - 2 - 3 152 95
DC325060.1 - 2 - 3 219 95

View Full Product Details