Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNMAL1
Entrez Gene ID
55228
Gene Name
paraneoplastic Ma antigen family like 1
Gene Aliases
-
Chromosome Location
Chr.19: 46466491 - 46471577 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018215.3 NP_060685.2 2 - 3 1607 98
GenBank mRNA AK001643.1 - 1 - 2 626 98
AK096326.1 - 2 - 3 1167 98
AK290218.1 - 3 - 4 1490 98
BC026026.1 - 1 - 2 640 98
BC032508.1 - 2 - 3 1585 98
BC051688.1 - 2 - 3 1564 98
BG708870.1 - 1 - 2 86 97

View Full Product Details